Please login to your online account to display your discounted pricing

2,3,6-Trifluorobenzoyl chloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3, 6-Trifluorobenzoyl chloride
CAS 189807-20-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H2ClF3O
Linear Formula F3C6HCOCl
Formula Weight 194.54
MDL Number MFCD00061203
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC312760010

  SDS
thermo fisher scientific
AC312760010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC312760050

  SDS
thermo fisher scientific
AC312760050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 194.54
Quantity 1g
Color Clear slightly yellow
Density 1.48g/mL
CAS Max % 100.0
Refractive Index 1.4815 to 1.4835 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Molecular Formula C7H2ClF3O
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 3, 6-Trifluorobenzoyl chloride
Assay Percent Range 98%
CAS 189807-20-3
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula F3C6HCOCl
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00061203
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.