Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,6-Trifluorobenzamide, 97%

$76.39 - $241.08

Chemical Identifiers

Chemical Name or Material 2,3,6-Trifluorobenzamide
CAS 207986-22-9
Molecular Formula C7H4F3NO
Formula Weight 175.11
MDL Number MFCD00061266
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1304303

  SDS
alfa aesar™
L1304303
1g Each for $76.39

AAL1304306

  SDS
alfa aesar™
L1304306
5g Each for $241.08
Description & Specifications

Specifications

Formula Weight 175.11
CAS 207986-22-9
MDL Number MFCD00061266
Assay 97%
Chemical Name or Material 2,3,6-Trifluorobenzamide
Molecular Formula C7H4F3NO
Health Hazard 1 Warning
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 115° to 118°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.