missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,6-Trifluorobenzamide, 97%

Manufacturer:  Alfa Aesar™ L1304306

 View more versions of this product

Catalog No. AAL1304306



Specifications

Specifications

2,3,6-Trifluorobenzamide
5g
C7H4F3NO
acmc-1cncg, benzamide,2,3,6-trifluoro, 2,3,6-tris fluoranyl benzamide
C1=CC(=C(C(=C1F)C(=O)N)F)F
175.11
175.11
115°C to 118°C
207986-22-9
MFCD00061266
BWWGEHSEZGXEKB-UHFFFAOYSA-N
2,3,6-trifluorobenzamide
594552
97%

Chemical Identifiers

207986-22-9
175.11
BWWGEHSEZGXEKB-UHFFFAOYSA-N
594552
C1=CC(=C(C(=C1F)C(=O)N)F)F
C7H4F3NO
MFCD00061266
acmc-1cncg, benzamide,2,3,6-trifluoro, 2,3,6-tris fluoranyl benzamide
2,3,6-trifluorobenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents