Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,6-Trifluorobenzaldehyde, 97%

$45.05 - $161.23

Chemical Identifiers

Chemical Name or Material 2,3,6-Trifluorobenzaldehyde
CAS 104451-70-9
Molecular Formula C7H3F3O
Formula Weight 160.1
MDL Number MFCD00061195
Beilstein 7701780
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2031903

  SDS
alfa aesar™
B2031903
1g Each for $45.05

AAB2031906

  SDS
alfa aesar™
B2031906
5g Each for $161.23
Description & Specifications

Specifications

Formula Weight 160.1
Chemical Name or Material 2,3,6-Trifluorobenzaldehyde
Assay 97%
Molecular Formula C7H3F3O
MDL Number MFCD00061195
CAS 104451-70-9
Beilstein 7701780
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 81°C (177°F)
Boiling Point 173°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.