Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,6-Trifluoroanisole, 97%

Chemical Identifiers

Chemical Name or Material 2,3,6-Trifluoroanisole
CAS 4920-34-7
Molecular Formula C7H5F3O
Formula Weight 162.11
MDL Number MFCD01631501
UN Number UN3271
Beilstein 2557137
Synonym 3-Methoxy-1,2,4-trifluorobenzene; 1-Methoxy-2,3,6-trifluorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2430203

  SDS
alfa aesar™
B2430203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2430206

  SDS
alfa aesar™
B2430206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.11
Quantity 1g
Molecular Formula C7H5F3O
Flash Point 45°C (113°F)
Beilstein 2557137
Synonym 3-Methoxy-1,2,4-trifluorobenzene; 1-Methoxy-2,3,6-trifluorobenzene
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
Chemical Name or Material 2,3,6-Trifluoroanisole
Health Hazard 1 Warning
Assay 97%
Boiling Point 146°C
UN Number UN3271
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD01631501
CAS 4920-34-7
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.