Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,6-Trifluoroaniline, 99%

Chemical Identifiers

Chemical Name or Material 2,3,6-Trifluoroaniline
CAS 67815-56-9
Molecular Formula C6H4F3N
Formula Weight 147.1
MDL Number MFCD00012369
UN Number UN2941
Beilstein 4976936
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2047503

 
alfa aesar™
B2047503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2047506

 
alfa aesar™
B2047506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.1
Chemical Name or Material 2,3,6-Trifluoroaniline
Assay 99%
Molecular Formula C6H4F3N
MDL Number MFCD00012369
CAS 67815-56-9
UN Number UN2941
Beilstein 4976936
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H373-H227-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P210-P260-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES
Flash Point 61°C (141°F)
Boiling Point 150° to 151°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P260-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H373-H227-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.