missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,6,7,10,11-Hexabromotriphenylene, TCI America™

$247.50 - $1,248.22

Chemical Identifiers

CAS 82632-80-2
Molecular Formula C18H6Br6
Molecular Weight (g/mol) 701.67
InChI Key GLHQUXLCQLQNPZ-UHFFFAOYSA-N
PubChem CID 11083045
IUPAC Name 2,3,6,7,10,11-hexabromotriphenylene
SMILES C1=C2C3=CC(=C(C=C3C4=CC(=C(C=C4C2=CC(=C1Br)Br)Br)Br)Br)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H1201100MG View Documents TCI America
H1201100MG
100mg Each for $247.50
 
H12011G View Documents TCI America
H12011G
1g Each for $1,248.22
 
Specifications

Chemical Identifiers

82632-80-2
701.67
11083045
C1=C2C3=CC(=C(C=C3C4=CC(=C(C=C4C2=CC(=C1Br)Br)Br)Br)Br)Br
C18H6Br6
GLHQUXLCQLQNPZ-UHFFFAOYSA-N
2,3,6,7,10,11-hexabromotriphenylene

Specifications

Undesignated
82632-80-2
GLHQUXLCQLQNPZ-UHFFFAOYSA-N
2,3,6,7,10,11-hexabromotriphenylene
11083045
Crystal-Powder at 20°C
2,3,6,7,10,11-Hexabromotriphenylene
100mg
C18H6Br6
C1=C2C3=CC(=C(C=C3C4=CC(=C(C=C4C2=CC(=C1Br)Br)Br)Br)Br)Br
701.67
701.67
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No