Learn More
2,3,5-Triphenyl-2H-tetrazolium chloride, 98%, Thermo Scientific Chemicals
2,3,5-Triphenyl-2H-tetrazolium chloride, 98%, C19H15ClN4, CAS Number-298-96-4, triphenyltetrazolium chloride, tetrazolium chloride, urocheck, tetrazolium red, tetrazolium chloride, uroscreen, red tetrazolium, vitastain, 2,3,5-triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride
$104.58 - $1336.37
Chemical Identifiers
CAS | 298-96-4 |
---|---|
Molecular Formula | C19H15ClN4 |
Molecular Weight (g/mol) | 334.81 |
MDL Number | MFCD00011963 |
InChI Key | PKDBCJSWQUOKDO-UHFFFAOYSA-M |
Synonym | 2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride |
PubChem CID | 9283 |
ChEBI | CHEBI:78019 |
IUPAC Name | 2,3,5-triphenyltetrazol-2-ium;chloride |
SMILES | [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1087009
|
Thermo Scientific Chemicals
A1087009 |
10 g |
Each for $104.58
|
|
|||||
AAA1087018
|
Thermo Scientific Chemicals
A1087018 |
50 g |
Each for $358.96
|
|
|||||
AAA1087030
|
Thermo Scientific Chemicals
A1087030 |
250 g |
Each for $1,336.37
|
|
|||||
Description
2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.
Solubility
Soluble in water, ethanol and acetone. Insoluble in ether.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Chemical Identifiers
298-96-4 | |
334.81 | |
PKDBCJSWQUOKDO-UHFFFAOYSA-M | |
9283 | |
2,3,5-triphenyltetrazol-2-ium;chloride |
C19H15ClN4 | |
MFCD00011963 | |
2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride | |
CHEBI:78019 | |
[Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
298-96-4 | |
98% | |
MFCD00011963 | |
3923356 | |
14,9744 | |
Soluble in water,ethanol and acetone. Insoluble in ether. | |
[Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1 | |
334.81 | |
CHEBI:78019 | |
98% |
∼250°C (decomposition) | |
C19H15ClN4 | |
10 g | |
Hygroscopic; Light sensitive | |
2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride | |
PKDBCJSWQUOKDO-UHFFFAOYSA-M | |
2,3,5-triphenyltetrazol-2-ium;chloride | |
9283 | |
334.81 | |
2,3,5-Triphenyl-2H-tetrazolium chloride |
Safety and Handling
GHS H Statement
H315-H319-H335
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-071-6
RTECSNumber : XF8100000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only