missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,5-Trimethylpyridine, TCI America™

Manufacturer:  TCI America T172225G

 View more versions of this product

Catalog No. T172225G



Specifications

Specifications

2,3,5-Trimethylpyridine
Yellow
695-98-7
MFCD00126840
GFYHSKONPJXCDE-UHFFFAOYSA-N
2,3,5-trimethylpyridine
12759
Clear Liquid at 20°C
65°C
25g
C8H11N
2,3,5-collidine, pyridine, 2,3,5-trimethyl, 2,3,5-trimethyl pyridine, 2,3,5-collidine, pubchem15901, acmc-1aul1, 2,3,5-trimethyl-pyridin, 2,3,5-trimethyl-pyridine, ksc490s0f, 2,3,5-collidine
CC1=CC(=C(N=C1)C)C
121.183
121.18
≥98.0% (GC)

Chemical Identifiers

695-98-7
121.183
GFYHSKONPJXCDE-UHFFFAOYSA-N
12759
CC1=CC(=C(N=C1)C)C
C8H11N
MFCD00126840
2,3,5-collidine, pyridine, 2,3,5-trimethyl, 2,3,5-trimethyl pyridine, 2,3,5-collidine, pubchem15901, acmc-1aul1, 2,3,5-trimethyl-pyridin, 2,3,5-trimethyl-pyridine, ksc490s0f, 2,3,5-collidine
2,3,5-trimethylpyridine
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0715

TSCA : No

Documents