Please login to your online account to display your discounted pricing

2,3,5-Trimethylpyrazine, 99%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  189970500

 View more versions of this product

Catalog No. AC189970500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 122.17
Quantity 50g
Color Clear yellow
RTECS Number UQ3907000
EINECS Number 238-712-0
TSCA TSCA
Density 0.97g/mL
Beilstein 23, 97
CAS Max % 100.0
Refractive Index 1.5030 to 1.5070 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Warning
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 3, 5-Trimethylpyrazine
Name Note 99%
Molecular Formula C7H10N2
CAS 14667-55-1
MDL Number MFCD00006145
Assay Percent Range 99%

Specifications

Chemical Name or Material 2, 3, 5-Trimethylpyrazine
Name Note 99%
CAS 14667-55-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H10N2
Formula Weight 122.17
MDL Number MFCD00006145
Accessories

GHS Signal Word: Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Flammable liquid and vapor.

SDS & Safety Documents