Please login to your online account to display your discounted pricing

2,3,5-Trimethylpyrazine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3, 5-Trimethylpyrazine
Name Note 99%
CAS 14667-55-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H10N2
Formula Weight 122.17
MDL Number MFCD00006145
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC189970500

  SDS
thermo fisher scientific
189970500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 122.17
Quantity 50g
Color Clear yellow
EINECS Number 238-712-0
TSCA TSCA
RTECS Number UQ3907000
Density 0.97g/mL
Beilstein 23, 97
CAS Max % 100.0
Chemical Name or Material 2, 3, 5-Trimethylpyrazine
Packaging Glass bottle
Refractive Index 1.5030 to 1.5070 (20°C, 589nm)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H10N2
MDL Number MFCD00006145
CAS 14667-55-1
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Flammable liquid and vapor.