missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5-Trimethylpyrazine, 99%

Manufacturer:  Alfa Aesar™ A1487030

 View more versions of this product

Catalog No. AAA1487030



Specifications

Specifications

2,3,5-Trimethylpyrazine
171°C to 172°C
14667-55-1
MFCD00006145
2423
IAEGWXHKWJGQAZ-UHFFFAOYSA-N
2,3,5-trimethylpyrazine
26808
99%
54°C (129°F)
250g
C7H10N2
UN1993
trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl
CC1=CN=C(C(=N1)C)C
122.171
122.17

Chemical Identifiers

14667-55-1
122.171
IAEGWXHKWJGQAZ-UHFFFAOYSA-N
26808
CC1=CN=C(C(=N1)C)C
C7H10N2
MFCD00006145
trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl
2,3,5-trimethylpyrazine
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : UQ3907000

TSCA : TSCA

SDS Documents