Please login to your online account to display your discounted pricing

2,3,5-Trimethylphenol, 98+%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  140190050

 View more versions of this product

Catalog No. AC140190050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Needle-Like Crystalline Powder
Formula Weight 136.19
Assay ≥98.0%
Quantity 5g
Color White to slightly yellow
EINECS Number 211-806-9
TSCA TSCA
Melting Point 92° to 95°C
CAS Min % 98.0
CAS Max % 100.0
Packaging Glass Bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Infrared Spectrum Authentic
Chemical Name or Material 2, 3, 5-Trimethylphenol
Name Note 98+%
Linear Formula (CH3)3C6H2OH
MDL Number MFCD00002228
Synonym Isopseudocumenol
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98+%
Molecular Formula C9H12O
CAS 697-82-5

Specifications

Chemical Name or Material 2, 3, 5-Trimethylphenol
Name Note 98+%
CAS 697-82-5
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C9H12O
Linear Formula (CH3)3C6H2OH
Formula Weight 136.19
MDL Number MFCD00002228
Synonym Isopseudocumenol
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents