missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,5-Trimethylanisole, TCI America™

Manufacturer:  TCI America T245625G

 View more versions of this product

Catalog No. T245625G



Specifications

Specifications

2,3,5-Trimethylanisole
Yellow
20469-61-8
MFCD00142783
AWONIZVBKXHWJP-UHFFFAOYSA-N
1-methoxy-2,3,5-trimethylbenzene
88555
Clear Liquid at 20°C
216°C
25g
C10H14O
2,3,5-trimethylanisole, anisole, 2,3,5-trimethyl, benzene, 1-methoxy-2,3,5-trimethyl, 1-methoxy-2,3,5-trimethyl-benzene, pubchem2688, acmc-209fam, 2,3,5-trimethylanizole, 3-methoxy-1,2,5-trimethylbenzene
CC1=CC(=C(C(=C1)OC)C)C
150.221
150.22
≥97.0% (GC)

Chemical Identifiers

20469-61-8
150.221
AWONIZVBKXHWJP-UHFFFAOYSA-N
88555
CC1=CC(=C(C(=C1)OC)C)C
C10H14O
MFCD00142783
2,3,5-trimethylanisole, anisole, 2,3,5-trimethyl, benzene, 1-methoxy-2,3,5-trimethyl, 1-methoxy-2,3,5-trimethyl-benzene, pubchem2688, acmc-209fam, 2,3,5-trimethylanizole, 3-methoxy-1,2,5-trimethylbenzene
1-methoxy-2,3,5-trimethylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents