missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triiodobenzoic acid, 98+%

CAS: 88-82-4 | C7H3I3O2 | 499.81 g/mol

$94.97 - $831.45

Chemical Identifiers

CAS 88-82-4
Molecular Formula C7H3I3O2
Molecular Weight (g/mol) 499.81
MDL Number MFCD00002420
InChI Key ZMZGFLUUZLELNE-UHFFFAOYSA-N
Synonym floraltone, tiba, triiodobenzoic acid, johnkolor, benzoic acid, 2,3,5-triiodo, 2,3,5-tiba, caswell no. 890a, triiodobenzoic acid van, regin 8, kyselina 2,3,5-trijodbenzoova
PubChem CID 6948
ChEBI CHEBI:73175
IUPAC Name 2,3,5-triiodobenzoic acid
SMILES OC(=O)C1=CC(I)=CC(I)=C1I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0267906
View Documents
Thermo Scientific Chemicals
L0267906
5 g
Each for $94.97
Only null left
 
AAL0267914
View Documents
Thermo Scientific Chemicals
L0267914
25 g
Each for $289.51
Only null left
 
AAL0267922
View Documents
Thermo Scientific Chemicals
L0267922
100 g
Each for $831.45
Only null left
 
Description

Description

2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents and light.
Specifications

Chemical Identifiers

88-82-4
499.81
ZMZGFLUUZLELNE-UHFFFAOYSA-N
6948
2,3,5-triiodobenzoic acid
C7H3I3O2
MFCD00002420
floraltone, tiba, triiodobenzoic acid, johnkolor, benzoic acid, 2,3,5-triiodo, 2,3,5-tiba, caswell no. 890a, triiodobenzoic acid van, regin 8, kyselina 2,3,5-trijodbenzoova
CHEBI:73175
OC(=O)C1=CC(I)=CC(I)=C1I

Specifications

88-82-4
C7H3I3O2
5 g
Light sensitive
Insoluble in water.
OC(=O)C1=CC(I)=CC(I)=C1I
499.81
CHEBI:73175
≥98%
220°C to 222°C
MFCD00002420
1955088
floraltone, tiba, triiodobenzoic acid, johnkolor, benzoic acid, 2,3,5-triiodo, 2,3,5-tiba, caswell no. 890a, triiodobenzoic acid van, regin 8, kyselina 2,3,5-trijodbenzoova
ZMZGFLUUZLELNE-UHFFFAOYSA-N
2,3,5-triiodobenzoic acid
6948
499.81
2,3,5-Triiodobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 201-859-6

RTECSNumber : DH9275000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only