missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,5-Trifluoropyridine, TCI America™

Manufacturer:  TCI America T277825G

 View more versions of this product

Catalog No. T277825G



Specifications

Specifications

2,3,5-Trifluoropyridine
Yellow
76469-41-5
MFCD03001162
pyridine, 2,3,5-trifluoro, pubchem8020, 2,3,5-trifluoro-pyridine, ksc494e2b, 2,3,5-tris fluoranyl pyridine, 2,3,5-trifluoropyridine 1g
C1=C(C=NC(=C1F)F)F
133.073
133.07
≥98.0% (GC)
102°C
25g
C5H2F3N
1993
JKVOXNTXYMXDHN-UHFFFAOYSA-N
2,3,5-trifluoropyridine
2783287
Clear Liquid at 20°C

Chemical Identifiers

76469-41-5
133.073
JKVOXNTXYMXDHN-UHFFFAOYSA-N
2783287
C1=C(C=NC(=C1F)F)F
C5H2F3N
MFCD03001162
pyridine, 2,3,5-trifluoro, pubchem8020, 2,3,5-trifluoro-pyridine, ksc494e2b, 2,3,5-tris fluoranyl pyridine, 2,3,5-trifluoropyridine 1g
2,3,5-trifluoropyridine
Safety and Handling

Safety and Handling

TSCA : No

Documents