Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5-Trifluoropyridine, 98+%

Chemical Identifiers

Chemical Name or Material 2,3,5-Trifluoropyridine
CAS 76469-41-5
Molecular Formula C5H2F3N
Formula Weight 133.07
MDL Number MFCD03001162
UN Number UN1993
Beilstein 6385503
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1954203

 
alfa aesar™
L1954203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1954206

 
alfa aesar™
L1954206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1954214

 
alfa aesar™
L1954214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 133.07
Assay ≥98%
Molecular Formula C5H2F3N
MDL Number MFCD03001162
CAS 76469-41-5
UN Number UN1993
Beilstein 6385503
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338-P337+P313
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Boiling Point 102°C
Chemical Name or Material 2,3,5-Trifluoropyridine
Flash Point 30°C (86°F)
Quantity 1g

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338-P337+P313
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.