Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5-Trifluoropyridine-4-carboxylic acid, 97%

$185.34 - $736.81

Chemical Identifiers

Chemical Name or Material 2,3,5-Trifluoropyridine-4-carboxylic acid
CAS 675602-91-2
Molecular Formula C6H2F3NO2
Formula Weight 177.08
MDL Number MFCD03840205
Synonym 2,3,5-Trifluoroisonicotinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2005003

  SDS
alfa aesar™
L2005003
1g Each for $185.34

AAL2005006

  SDS
alfa aesar™
L2005006
5g Each for $736.81
Description & Specifications

Specifications

Formula Weight 177.08
Molecular Formula C6H2F3NO2
MDL Number MFCD03840205
Chemical Name or Material 2,3,5-Trifluoropyridine-4-carboxylic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
CAS 675602-91-2
Quantity 1g
Health Hazard 1 Warning
Synonym 2,3,5-Trifluoroisonicotinic acid
Melting Point 108° to 112°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.