missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,5-Trifluorobenzoic Acid 98.0+%, TCI America™

$149.32 - $593.92

Chemical Identifiers

CAS 654-87-5
Molecular Formula C7H3F3O2
Molecular Weight (g/mol) 176.09
MDL Number MFCD00061200
InChI Key CPZROMDDCPPFOO-UHFFFAOYSA-N
Synonym 2,3,5-trilfluorobenzoic acid, benzoic acid, 2,3,5-trifluoro, pubchem1401, acmc-209nr1, 2,3,5-trifluorobenzoicacid, ksc622o6j, 2,3,5-trifluoro-benzoic acid, rarechem al bo 0710, benzoic acid,2,3,5-trifluoro, cpzromddcppfoo-uhfffaoysa
PubChem CID 2776991
IUPAC Name 2,3,5-trifluorobenzoic acid
SMILES OC(=O)C1=CC(F)=CC(F)=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T23851G
View Documents Promotion Details
TCI America
T23851G
1 g
Each for $149.32
 
T23855G
View Documents Promotion Details
TCI America
T23855G
5 g
Each for $593.92
 
Description

Description

Specifications

Chemical Identifiers

654-87-5
176.09
CPZROMDDCPPFOO-UHFFFAOYSA-N
2776991
OC(=O)C1=CC(F)=CC(F)=C1F
C7H3F3O2
MFCD00061200
2,3,5-trilfluorobenzoic acid, benzoic acid, 2,3,5-trifluoro, pubchem1401, acmc-209nr1, 2,3,5-trifluorobenzoicacid, ksc622o6j, 2,3,5-trifluoro-benzoic acid, rarechem al bo 0710, benzoic acid,2,3,5-trifluoro, cpzromddcppfoo-uhfffaoysa
2,3,5-trifluorobenzoic acid

Specifications

654-87-5
White-Yellow
MFCD00061200
2,3,5-trilfluorobenzoic acid, benzoic acid, 2,3,5-trifluoro, pubchem1401, acmc-209nr1, 2,3,5-trifluorobenzoicacid, ksc622o6j, 2,3,5-trifluoro-benzoic acid, rarechem al bo 0710, benzoic acid,2,3,5-trifluoro, cpzromddcppfoo-uhfffaoysa
OC(=O)C1=CC(F)=CC(F)=C1F
176.09
176.09
Crystalline Powder
109°C
C7H3F3O2
1 g
CPZROMDDCPPFOO-UHFFFAOYSA-N
2,3,5-trifluorobenzoic acid
2776991
≥98.0% (GC,T)
2,3,5-Trifluorobenzoic Acid
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'