Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5-Trifluorobenzenesulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2,3,5-Trifluorobenzenesulfonyl chloride
CAS 914636-99-0
Molecular Formula C6H2ClF3O2S
Formula Weight 230.59
MDL Number MFCD08532462
UN Number UN3261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3233103

  SDS
alfa aesar™
H3233103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH32331MD

  SDS
alfa aesar™
H32331MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.59
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C6H2ClF3O2S
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Assay 97%
Melting Point 40° to 49°C
MDL Number MFCD08532462
Chemical Name or Material 2,3,5-Trifluorobenzenesulfonyl chloride
UN Number UN3261
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Quantity 1g
CAS 914636-99-0
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.