Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5-Trifluorobenzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 2,3,5-Trifluorobenzaldehyde
CAS 126202-23-1
Molecular Formula C7H3F3O
Formula Weight 160.1
MDL Number MFCD00083538
UN Number UN1989
Beilstein 9190838
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2034003

  SDS
alfa aesar™
B2034003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2034006

  SDS
alfa aesar™
B2034006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.1
Chemical Name or Material 2,3,5-Trifluorobenzaldehyde
Molecular Formula C7H3F3O
CAS 126202-23-1
Assay 97%
Health Hazard 1 Warning
Beilstein 9190838
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Boiling Point 165°C
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
UN Number UN1989
Flash Point 57°C (135°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
MDL Number MFCD00083538
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.