Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5-Trifluoroaniline, 97%

Chemical Identifiers

Chemical Name or Material 2,3,5-Trifluoroaniline
CAS 363-80-4
Molecular Formula C6H4F3N
Formula Weight 147.1
MDL Number MFCD00083549
UN Number UN2941
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2683103

  SDS
alfa aesar™
H2683103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH26831MD

  SDS
alfa aesar™
H26831MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.1
Chemical Name or Material 2,3,5-Trifluoroaniline
Assay 97%
Molecular Formula C6H4F3N
MDL Number MFCD00083549
CAS 363-80-4
UN Number UN2941
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.