Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',3',5'-Trifluoroacetophenone, 97%

$130.19 - $394.77

Chemical Identifiers

Chemical Name or Material 2',3',5'-Trifluoroacetophenone
CAS 243459-93-0
Molecular Formula C8H5F3O
Formula Weight 174.12
MDL Number MFCD00083548
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2682803

  SDS
alfa aesar™
H2682803
1g Each for $130.19

AAH2682806

  SDS
alfa aesar™
H2682806
5g Each for $394.77
Description & Specifications

Specifications

Formula Weight 174.12
Chemical Name or Material 2',3',5'-Trifluoroacetophenone
CAS 243459-93-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Quantity 1g
MDL Number MFCD00083548
Molecular Formula C8H5F3O
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.