missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5-Trichlorothiophene, 97%

$45.04 - $111.18

Chemical Identifiers

CAS 17249-77-3
Molecular Formula C4HCl3S
Molecular Weight (g/mol) 187.462
MDL Number MFCD01318176
InChI Key AZIFHMYTYFPVMV-UHFFFAOYSA-N
Synonym thiophene, 2,3,5-trichloro, 2,3,5-trichloro-thiophene, acmc-1ch2b, thiophene,2,3,5-trichloro
PubChem CID 123374
IUPAC Name 2,3,5-trichlorothiophene
SMILES C1=C(SC(=C1Cl)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1720703 View Documents Alfa Aesar™
L1720703
1g Each for $111.18
 
AAL17207MD View Documents Alfa Aesar™
L17207MD
250mg Each for $45.04
 
Specifications

Chemical Identifiers

17249-77-3
187.462
AZIFHMYTYFPVMV-UHFFFAOYSA-N
123374
C1=C(SC(=C1Cl)Cl)Cl
C4HCl3S
MFCD01318176
thiophene, 2,3,5-trichloro, 2,3,5-trichloro-thiophene, acmc-1ch2b, thiophene,2,3,5-trichloro
2,3,5-trichlorothiophene

Specifications

110°C (20mmHg)
1g
C4HCl3S
UN2810
thiophene, 2,3,5-trichloro, 2,3,5-trichloro-thiophene, acmc-1ch2b, thiophene,2,3,5-trichloro
C1=C(SC(=C1Cl)Cl)Cl
187.462
187.48
2,3,5-Trichlorothiophene
-16°C
17249-77-3
MFCD01318176
2431
AZIFHMYTYFPVMV-UHFFFAOYSA-N
2,3,5-trichlorothiophene
123374
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.