2,3,5-Trichloropyridine, ACROS Organics™
Manufacturer:  Thermo Fisher Scientific AC247020500
 View more versions of this product
Catalog No. AC247020500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Specifications
2, 3, 5-Trichloropyridine
Glass bottle
46°C to 50°C
16063-70-0
MFCD00043007
CNLIIAKAAMFCJG-UHFFFAOYSA-N
2,3,5-trichloropyridine
27666
113°C
219°C
50g
C5H2Cl3N
2, 3, 5-trichloropyridine, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, 2,3,6-trichloro-pyridine, acmc-1at5w, g00013-watson-int, ksc179q6d, pubchem1127, pyridine, 2,3,5-trichloro, tpc-i005
C1=C(C=NC(=C1Cl)Cl)Cl
182.44
182.44
Chemical Identifiers
16063-70-0
182.44
CNLIIAKAAMFCJG-UHFFFAOYSA-N
27666
C1=C(C=NC(=C1Cl)Cl)Cl
C5H2Cl3N
MFCD00043007
2, 3, 5-trichloropyridine, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, 2,3,6-trichloro-pyridine, acmc-1at5w, g00013-watson-int, ksc179q6d, pubchem1127, pyridine, 2,3,5-trichloro, tpc-i005
2,3,5-trichloropyridine
Safety and Handling
Safety and Handling
GHS H Statement:
Harmful to aquatic life with long lasting effects.
GHS P Statement:
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS Documents