missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,5-Trichloropyridine, ≥98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6417322

 View more versions of this product

Catalog No. AAH6417322



Description

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications

Specifications

2,3,5-Trichloropyridine
46°C to 50°C (literature)
219°C (literature)
C5H2Cl3N
100 g
pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005
CNLIIAKAAMFCJG-UHFFFAOYSA-N
2,3,5-trichloropyridine
27666
≥98%
16063-70-0
White to Yellow
113°C (235°F)
MFCD00043007
119384
Slightly soluble in water.
C1=C(C=NC(=C1Cl)Cl)Cl
182.428
182.43
Solid

Chemical Identifiers

16063-70-0
182.428
CNLIIAKAAMFCJG-UHFFFAOYSA-N
27666
C1=C(C=NC(=C1Cl)Cl)Cl
C5H2Cl3N
MFCD00043007
pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005
2,3,5-trichloropyridine
Safety and Handling

Safety and Handling

EINECSNumber : 407-270-2

RTECSNumber : UU0525000

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only