missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5-Trichloropyridine-4-carboxylic acid, 95%

Manufacturer:  Alfa Aesar™ H6358806

 View more versions of this product

Catalog No. AAH6358806



Specifications

Specifications

2,3,5-Trichloropyridine-4-carboxylic acid
5g
C6H2Cl3NO2
2,3,5-trichloroisonicotinic acid, 2,3,5-trichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,3,5-trichloro, trichloroisonicotinicacid, acmc-1bmtq, ksc235e5t, 2,3,5-trichloroisonicotinicacid, 2,3,5-trichloro-isonicotinic acid, 4-carboxy-2,3,5-trichloropyridine, 2,3,5-trichloropyridine-4-carboxylicacid
C1=C(C(=C(C(=N1)Cl)Cl)C(=O)O)Cl
226.437
226.44
95%
White
406676-18-4
MFCD11044390
TYMTYHMQJHSRIF-UHFFFAOYSA-N
2,3,5-trichloropyridine-4-carboxylic acid
10944163
Powder

Chemical Identifiers

406676-18-4
226.437
TYMTYHMQJHSRIF-UHFFFAOYSA-N
10944163
C1=C(C(=C(C(=N1)Cl)Cl)C(=O)O)Cl
C6H2Cl3NO2
MFCD11044390
2,3,5-trichloroisonicotinic acid, 2,3,5-trichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,3,5-trichloro, trichloroisonicotinicacid, acmc-1bmtq, ksc235e5t, 2,3,5-trichloroisonicotinicacid, 2,3,5-trichloro-isonicotinic acid, 4-carboxy-2,3,5-trichloropyridine, 2,3,5-trichloropyridine-4-carboxylicacid
2,3,5-trichloropyridine-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents