missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,5-Trichlorobenzaldehyde, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC243530100

 View more versions of this product

Catalog No. AC243530100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 3, 5-Trichlorobenzaldehyde
Authentic
White or Yellow
10g
96.0
96% min. (GC)
MFCD00010860
2,3,5-trichlorobenzaldehyde, acmc-1b126, benzaldehyde, 2,3,5-trichloro, benzaldehyde,2,3,5-trichloro, djyrztclvdkwbl-uhfffaoysa, ksc606a4b
C1=C(C=C(C(=C1Cl)Cl)C=O)Cl
209.46
209.46
97%
>110°C
Glass bottle
72°C to 76°C
56961-75-2
100.0
C7H3Cl3O
07, 180
DJYRZTCLVDKWBL-UHFFFAOYSA-N
2,3,5-trichlorobenzaldehyde
92596
Powder and/or Chunks

Chemical Identifiers

56961-75-2
209.46
DJYRZTCLVDKWBL-UHFFFAOYSA-N
92596
C1=C(C=C(C(=C1Cl)Cl)C=O)Cl
C7H3Cl3O
MFCD00010860
2,3,5-trichlorobenzaldehyde, acmc-1b126, benzaldehyde, 2,3,5-trichloro, benzaldehyde,2,3,5-trichloro, djyrztclvdkwbl-uhfffaoysa, ksc606a4b
2,3,5-trichlorobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents