missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,5-Tribromothiophene 98.0+%, TCI America™

$141.41 - $742.36

Chemical Identifiers

CAS 3141-24-0
Molecular Formula C4HBr3S
Molecular Weight (g/mol) 320.82
MDL Number MFCD00014521
InChI Key SKDNDSLDRLEELJ-UHFFFAOYSA-N
Synonym 2,3,5-tribromo thiophene, 2,3,5-tribromothiophene, 2,5-tribromothiophene, acmc-209hmg, ksc225k9l, pubchem5608, thiophene, 2,3,5-tribromo, thiophene,3,5-tribromo
PubChem CID 76592
IUPAC Name 2,3,5-tribromothiophene
SMILES BrC1=CC(Br)=C(Br)S1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T095625G
View Documents
Tci America
T095625G
25g
Each for $141.41
 
T0956250G
View Documents
Tci America
T0956250G
250g
Each for $742.36
 
Description

Description

Specifications

Chemical Identifiers

3141-24-0
320.82
SKDNDSLDRLEELJ-UHFFFAOYSA-N
76592
BrC1=CC(Br)=C(Br)S1
C4HBr3S
MFCD00014521
2,3,5-tribromo thiophene, 2,3,5-tribromothiophene, 2,5-tribromothiophene, acmc-209hmg, ksc225k9l, pubchem5608, thiophene, 2,3,5-tribromo, thiophene,3,5-tribromo
2,3,5-tribromothiophene

Specifications

Yellow
3141-24-0
MFCD00014521
SKDNDSLDRLEELJ-UHFFFAOYSA-N
2,3,5-tribromothiophene
76592
Crystal-Lump at 20°C
2,3,5-Tribromothiophene
25g
C4HBr3S
2,3,5-tribromo thiophene, 2,3,5-tribromothiophene, 2,5-tribromothiophene, acmc-209hmg, ksc225k9l, pubchem5608, thiophene, 2,3,5-tribromo, thiophene,3,5-tribromo
BrC1=CC(Br)=C(Br)S1
320.82
320.82
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

SDS