Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5-Tribromothiophene, 98%

Chemical Identifiers

Chemical Name or Material 2,3,5-Tribromothiophene
CAS 3141-24-0
Molecular Formula C4HBr3S
Formula Weight 320.84
MDL Number MFCD00014521
UN Number UN2811
Beilstein 111486
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1176106

  SDS
alfa aesar™
A1176106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1176114

  SDS
alfa aesar™
A1176114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1176122

  SDS
alfa aesar™
A1176122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 320.84
UN Number UN2811
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C4HBr3S
Boiling Point 77° to 78°C (0.7mmHg)
Chemical Name or Material 2,3,5-Tribromothiophene
MDL Number MFCD00014521
Health Hazard 1 Danger
Beilstein 111486
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 25° to 29°C
CAS 3141-24-0
TSCA TSCA
Flash Point >110°C (230°F)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 5g
Assay 98%
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.