missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5-Tribromothiophene, 98%

Manufacturer:  Alfa Aesar™ A1176106

 View more versions of this product

Catalog No. AAA1176106



Specifications

Specifications

2,3,5-Tribromothiophene
77°C to 78°C (0.7mmHg)
5g
C4HBr3S
UN2811
thiophene, 2,3,5-tribromo, pubchem5608, 2,5-tribromothiophene, acmc-209hmg, thiophene,3,5-tribromo, 2,3,5-tribromo thiophene, ksc225k9l, 2,3,5-tribromothiophene
C1=C(SC(=C1Br)Br)Br
320.824
320.84
>110°C (230°F)
25°C to 29°C
3141-24-0
MFCD00014521
111486
SKDNDSLDRLEELJ-UHFFFAOYSA-N
2,3,5-tribromothiophene
76592
98%

Chemical Identifiers

3141-24-0
320.824
SKDNDSLDRLEELJ-UHFFFAOYSA-N
76592
C1=C(SC(=C1Br)Br)Br
C4HBr3S
MFCD00014521
thiophene, 2,3,5-tribromo, pubchem5608, 2,5-tribromothiophene, acmc-209hmg, thiophene,3,5-tribromo, 2,3,5-tribromo thiophene, ksc225k9l, 2,3,5-tribromothiophene
2,3,5-tribromothiophene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents