missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5-Tri-O-benzyl-L-arabinofuranose, 98%

$45.18 - $348.51

Chemical Identifiers

CAS 89615-42-9
Molecular Formula C26H28O5
Molecular Weight (g/mol) 420.505
MDL Number MFCD00053542
InChI Key NAQUAXSCBJPECG-SSUZURRFSA-N
Synonym 2,3,5-tri-o-benzyl-beta-l-arabinofuranose, 2s,3r,4s,5s-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol, 2,3,5-tri-o-benzyl-l-arabinofuranose, 2s,3r,4s,5s-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, 2,3,5-tri-o-benzyl-beta-l-arabino-furanose, b-l-arabinofuranose,2,3,5-tris-o-phenylmethyl
PubChem CID 12933788
IUPAC Name (2S,3R,4S,5S)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
SMILES C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66343MD View Documents Alfa Aesar™
J66343MD
250mg Each for $45.18
 
AAJ6634303 View Documents Alfa Aesar™
J6634303
1g Each for $119.20
 
AAJ6634306 View Documents Alfa Aesar™
J6634306
5g Each for $348.51
 
Specifications

Chemical Identifiers

89615-42-9
420.505
NAQUAXSCBJPECG-SSUZURRFSA-N
12933788
C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4
C26H28O5
MFCD00053542
2,3,5-tri-o-benzyl-beta-l-arabinofuranose, 2s,3r,4s,5s-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol, 2,3,5-tri-o-benzyl-l-arabinofuranose, 2s,3r,4s,5s-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, 2,3,5-tri-o-benzyl-beta-l-arabino-furanose, b-l-arabinofuranose,2,3,5-tris-o-phenylmethyl
(2S,3R,4S,5S)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol

Specifications

White
89615-42-9
MFCD00053542
NAQUAXSCBJPECG-SSUZURRFSA-N
(2S,3R,4S,5S)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
12933788
Powder
2,3,5-Tri-O-benzyl-L-arabinofuranose
250mg
C26H28O5
2,3,5-tri-o-benzyl-beta-l-arabinofuranose, 2s,3r,4s,5s-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol, 2,3,5-tri-o-benzyl-l-arabinofuranose, 2s,3r,4s,5s-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, 2,3,5-tri-o-benzyl-beta-l-arabino-furanose, b-l-arabinofuranose,2,3,5-tris-o-phenylmethyl
C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4
420.505
420.5
98%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C