missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5-Tri-O-benzyl-beta-D-ribofuranose, 98%

$60.34 - $113.14

Chemical Identifiers

CAS 89361-52-4
Molecular Formula C26H28O5
Formula Weight 420.5
MDL Number MFCD01864312
Chemical Name or Material 2,3,5-Tri-O-benzyl-beta-D-ribofuranose
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6625204 SDS Alfa Aesar™
J6625204
2g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ6625206 SDS Alfa Aesar™
J6625206
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ6625203 SDS Alfa Aesar™
J6625203
1g Each for $113.14
AAJ66252MD SDS Alfa Aesar™
J66252MD
250mg Each for $60.34
Specifications

Description

Solubility
Soluble in chloroform

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Description & Specifications

Chemical Identifiers

89361-52-4
420.5
2,3,5-Tri-O-benzyl-beta-D-ribofuranose
C26H28O5
MFCD01864312

Specifications

+43° (c=2 in Dioxane)
2g
Powder
420.5
Soluble in chloroform
98%
White
89361-52-4
C26H28O5
MFCD01864312
2,3,5-Tri-O-benzyl-beta-D-ribofuranose