missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoic acid, 97%, Maybridge

Manufacturer:  Thermo Scientific CC15701CB

 View more versions of this product

Catalog No. CC15701CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(3, 5-Dimethyl-1H-pyrazol-4-yl)benzoic acid
250mg
96.5
97%
MFCD02682016
JNHKHFADRGJMJS-UHFFFAOYSA-N
2-(3,5-dimethyl-1H-pyrazol-4-yl)benzoic acid
2776446
97%
Amber Glass Bottle
321309-43-7
100.0
C12H12N2O2
2-3,5-dimethyl-1h-pyrazol-4-yl benzoic acid, 4-2-carboxyphenyl-3,5-dimethylpyrazole, benzoic acid,2-3,5-dimethyl-1h-pyrazol-4-yl, 2-3,5-dimethylpyrazol-4-yl benzoic acid, 2-3,5-dimethyl-1h-pyrazol-4-yl-benzoic acid
CC1=C(C(=NN1)C)C2=CC=CC=C2C(=O)O
216.24
216.24

Chemical Identifiers

321309-43-7
216.24
JNHKHFADRGJMJS-UHFFFAOYSA-N
2776446
CC1=C(C(=NN1)C)C2=CC=CC=C2C(=O)O
C12H12N2O2
MFCD02682016
2-3,5-dimethyl-1h-pyrazol-4-yl benzoic acid, 4-2-carboxyphenyl-3,5-dimethylpyrazole, benzoic acid,2-3,5-dimethyl-1h-pyrazol-4-yl, 2-3,5-dimethylpyrazol-4-yl benzoic acid, 2-3,5-dimethyl-1h-pyrazol-4-yl-benzoic acid
2-(3,5-dimethyl-1H-pyrazol-4-yl)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents