Please login to your online account to display your discounted pricing

2-[3,5-di(tert-butyl)-4-hydroxyphenyl]acetic acid, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  BTB09885EA

 View more versions of this product

Catalog No. BTB09885EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 264.36
Quantity 10g
Assay 97%
Chemical Name or Material 2-[3, 5-di(tert-butyl)-4-hydroxyphenyl]acetic acid
Molecular Formula C16H24O3
CAS 1611-03-6

Specifications

Chemical Name or Material 2-[3, 5-di(tert-butyl)-4-hydroxyphenyl]acetic acid
Assay 97%
Molecular Formula C16H24O3
Formula Weight 264.36