missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(3,5-Di-tert-amyl-2-hydroxyphenyl)benzotriazole, TCI America™

$41.25 - $359.03

Chemical Identifiers

CAS 25973-55-1
Molecular Formula C22H29N3O
Molecular Weight (g/mol) 351.494
MDL Number MFCD00142737
InChI Key ZMWRRFHBXARRRT-UHFFFAOYSA-N
PubChem CID 33263
IUPAC Name 2-(benzotriazol-2-yl)-4,6-bis(2-methylbutan-2-yl)phenol
SMILES CCC(C)(C)C1=CC(=C(C(=C1)N2N=C3C=CC=CC3=N2)O)C(C)(C)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H071825G View Documents TCI America
H071825G
25g Each for $41.25
 
H0718500G View Documents TCI America
H0718500G
500g Each for $359.03
 
Specifications

Chemical Identifiers

25973-55-1
351.494
ZMWRRFHBXARRRT-UHFFFAOYSA-N
2-(benzotriazol-2-yl)-4,6-bis(2-methylbutan-2-yl)phenol
C22H29N3O
MFCD00142737
33263
CCC(C)(C)C1=CC(=C(C(=C1)N2N=C3C=CC=CC3=N2)O)C(C)(C)CC

Specifications

Yellow
25g
C22H29N3O
ZMWRRFHBXARRRT-UHFFFAOYSA-N
2-(benzotriazol-2-yl)-4,6-bis(2-methylbutan-2-yl)phenol
33263
Crystal-Powder at 20°C
2-(3,5-Di-tert-amyl-2-hydroxyphenyl)benzotriazole
82°C
25973-55-1
MFCD00142737
CCC(C)(C)C1=CC(=C(C(=C1)N2N=C3C=CC=CC3=N2)O)C(C)(C)CC
351.494
351.49
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3604

TSCA : Yes