Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-[3,5-Bis(trifluoromethyl)phenyl]thioacetamide, 97%

$171.73 - $171.73

Chemical Identifiers

Chemical Name or Material 2-[3,5-Bis(trifluoromethyl)phenyl]thioacetamide
CAS 691868-49-2
Molecular Formula C10H7F6NS
Formula Weight 287.22g/mol
MDL Number MFCD01313627
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2654203

 
alfa aesar™
H26542-03
1g Each for $171.73
Description & Specifications

Specifications

Melting Point 109° to 111°C
Quantity 1g
Chemical Name or Material 2-[3,5-Bis(trifluoromethyl)phenyl]thioacetamide
CAS 691868-49-2
Assay 97%
Molecular Formula C10H7F6NS
Formula Weight 287.22g/mol
MDL Number MFCD01313627
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.