missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5,6-Tetramethylbenzyl chloride, 98+%

$95.92 - $388.61

Chemical Identifiers

CAS 7435-83-8
Molecular Formula C11H15Cl
Molecular Weight (g/mol) 182.691
MDL Number MFCD00013679
InChI Key UGAPPXGBBWAIGT-UHFFFAOYSA-N
Synonym 3-chloromethyl-1,2,4,5-tetramethylbenzene, 2,3,5,6-tetramethylbenzyl chloride, benzene, 3-chloromethyl-1,2,4,5-tetramethyl, 4-05-00-01111 beilstein handbook reference, 2,3,5,6-tetramethylbenzylchloride, 2,3,5,6-tetramethyl benzyl chloride, 2,3,5,6-tetramethyl-benzyl chloride, 3-chloromethyl-1,2,4,5-tetramethyl-benzene, 3-chloromethyl-1,2,4,5-tetramethyl-benzene, benzene,3-chloromethyl-1,2,4,5-tetramethyl
PubChem CID 81939
IUPAC Name 3-(chloromethyl)-1,2,4,5-tetramethylbenzene
SMILES CC1=CC(=C(C(=C1C)CCl)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0050106 View Documents Alfa Aesar™
L0050106
5g Each for $95.92
 
AAL0050114 View Documents Alfa Aesar™
L0050114
25g Each for $388.61
 
Specifications

Chemical Identifiers

7435-83-8
182.691
UGAPPXGBBWAIGT-UHFFFAOYSA-N
81939
CC1=CC(=C(C(=C1C)CCl)C)C
C11H15Cl
MFCD00013679
3-chloromethyl-1,2,4,5-tetramethylbenzene, 2,3,5,6-tetramethylbenzyl chloride, benzene, 3-chloromethyl-1,2,4,5-tetramethyl, 4-05-00-01111 beilstein handbook reference, 2,3,5,6-tetramethylbenzylchloride, 2,3,5,6-tetramethyl benzyl chloride, 2,3,5,6-tetramethyl-benzyl chloride, 3-chloromethyl-1,2,4,5-tetramethyl-benzene, 3-chloromethyl-1,2,4,5-tetramethyl-benzene, benzene,3-chloromethyl-1,2,4,5-tetramethyl
3-(chloromethyl)-1,2,4,5-tetramethylbenzene

Specifications

68°C to 71°C
7435-83-8
MFCD00013679
1863560
UGAPPXGBBWAIGT-UHFFFAOYSA-N
3-(chloromethyl)-1,2,4,5-tetramethylbenzene
81939
≥98%
5g
C11H15Cl
UN3261
3-chloromethyl-1,2,4,5-tetramethylbenzene, 2,3,5,6-tetramethylbenzyl chloride, benzene, 3-chloromethyl-1,2,4,5-tetramethyl, 4-05-00-01111 beilstein handbook reference, 2,3,5,6-tetramethylbenzylchloride, 2,3,5,6-tetramethyl benzyl chloride, 2,3,5,6-tetramethyl-benzyl chloride, 3-chloromethyl-1,2,4,5-tetramethyl-benzene, 3-chloromethyl-1,2,4,5-tetramethyl-benzene, benzene,3-chloromethyl-1,2,4,5-tetramethyl
CC1=CC(=C(C(=C1C)CCl)C)C
182.691
182.69
2,3,5,6-Tetramethylbenzyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : CZ0825000