Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5,6-Tetramethylbenzyl alcohol, 98+%

$32.26 - $101.57

Chemical Identifiers

Chemical Name or Material 2,3,5,6-Tetramethylbenzyl alcohol
CAS 78985-13-4
Molecular Formula C11H16O
Formula Weight 164.25
MDL Number MFCD00051485
Beilstein 4968835
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0238403

  SDS
alfa aesar™
L0238403
1g Each for $32.26

AAL0238406

  SDS
alfa aesar™
L0238406
5g Each for $101.57
Description & Specifications

Specifications

Formula Weight 164.25
Chemical Name or Material 2,3,5,6-Tetramethylbenzyl alcohol
Assay ≥98%
Molecular Formula C11H16O
MDL Number MFCD00051485
CAS 78985-13-4
Beilstein 4968835
Melting Point 110° to 114°C
Quantity 1g