Please login to your online account to display your discounted pricing

2,3,5,6-Tetramethylbenzenesulfonyl chloride, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC356950250

 View more versions of this product

Catalog No. AC356950250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (ex Cl)
Physical Form Crystalline Powder
Formula Weight 232.73
Quantity 25g
Color White to pale cream
Melting Point 97° to 101°C
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
CAS 60706-63-0
Assay Percent Range 98%
Molecular Formula C10H13ClO2S
Chemical Name or Material 2,3,5,6-Tetramethylbenzenesulfonyl chloride
Name Note 98%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
CAS Min % 97.5
Packaging Glass bottle
MDL Number MFCD00014723
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2,3,5,6-Tetramethylbenzenesulfonyl chloride
Name Note 98%
CAS 60706-63-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (ex Cl)
Assay Percent Range 98%
Molecular Formula C10H13ClO2S
Formula Weight 232.73
MDL Number MFCD00014723
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents