Please login to your online account to display your discounted pricing

2,3,5,6-Tetramethylbenzenesulfonyl chloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,3,5,6-Tetramethylbenzenesulfonyl chloride
Name Note 98%
CAS 60706-63-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (ex Cl)
Assay Percent Range 98%
Molecular Formula C10H13ClO2S
Formula Weight 232.73
MDL Number MFCD00014723
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC356950250

  SDS
thermo fisher scientific
AC356950250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC356950100

  SDS
thermo fisher scientific
AC356950100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (ex Cl)
Physical Form Crystalline Powder
Formula Weight 232.73
Quantity 25g
Color White to pale cream
Melting Point 97° to 101°C
CAS Max % 100.0
Chemical Name or Material 2,3,5,6-Tetramethylbenzenesulfonyl chloride
Infrared Spectrum Authentic
Name Note 98%
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C10H13ClO2S
MDL Number MFCD00014723
CAS 60706-63-0
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.