Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',3',5',6'-Tetramethylacetophenone, 98+%

$48.34 - $164.70

Chemical Identifiers

Chemical Name or Material 2',3',5',6'-Tetramethylacetophenone
CAS 2142-79-2
Molecular Formula C12H16O
Formula Weight 176.26
MDL Number MFCD00014987
Beilstein 2087755
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0107006

 
alfa aesar™
L0107006
5g Each for $48.34

AAL0107014

 
alfa aesar™
L0107014
25g Each for $164.70
Description & Specifications

Specifications

Formula Weight 176.26
Chemical Name or Material 2',3',5',6'-Tetramethylacetophenone
CAS 2142-79-2
Assay ≥98%
Molecular Formula C12H16O
MDL Number MFCD00014987
Beilstein 2087755
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Melting Point 74° to 75°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.