Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5,6-Tetrafluoropyridine-4-propionic acid, 98%

$151.78 - $151.78

Chemical Identifiers

Chemical Name or Material 2,3,5,6-Tetrafluoropyridine-4-propionic acid
CAS 916792-08-0
Molecular Formula C8H5F4NO2
Formula Weight 223.13g/mol
MDL Number MFCD08064226
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25768MD

 
alfa aesar™
H25768-MD
250mg Each for $151.78
Description & Specifications

Specifications

Melting Point 151° to 153°C
Quantity 250mg
Chemical Name or Material 2,3,5,6-Tetrafluoropyridine-4-propionic acid
CAS 916792-08-0
Assay 98%
Molecular Formula C8H5F4NO2
Formula Weight 223.13g/mol
MDL Number MFCD08064226
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.