Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5,6-Tetrafluorophenylhydrazine, 97%

$66.15 - $192.86

Chemical Identifiers

Chemical Name or Material 2,3,5,6-Tetrafluorophenylhydrazine
CAS 653-11-2
Molecular Formula C6H4F4N2
Formula Weight 180.11
MDL Number MFCD00007575
Beilstein 745777
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2260006

  SDS
alfa aesar™
B2260006
5g Each for $66.15

AAB2260014

  SDS
alfa aesar™
B2260014
25g Each for $192.86
Description & Specifications

Specifications

Formula Weight 180.11
Chemical Name or Material 2,3,5,6-Tetrafluorophenylhydrazine
Assay 97%
Molecular Formula C6H4F4N2
MDL Number MFCD00007575
CAS 653-11-2
Beilstein 745777
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 86° to 92°C
Quantity 5g
Recommended Storage 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C