Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5,6-Tetrafluorophenol, 97%

Chemical Identifiers

Chemical Name or Material 2,3,5,6-Tetrafluorophenol
CAS 769-39-1
Molecular Formula C6H2F4O
Formula Weight 166.07
MDL Number MFCD00002157
UN Number UN3261
Beilstein 1911548
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1268606

  SDS
alfa aesar™
A1268606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1268614

  SDS
alfa aesar™
A1268614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.07
Quantity 5g
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 79°C (174°F)
Molecular Formula C6H2F4O
Beilstein 1911548
Boiling Point 141° to 143°C
Chemical Name or Material 2,3,5,6-Tetrafluorophenol
MDL Number MFCD00002157
UN Number UN3261
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
CAS 769-39-1
Health Hazard 2 GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Odor Phenol-like
Assay 97%
Melting Point 28° to 30°C

Applications
2,3,5,6-Tetrafluorophenol is used as a cytochrome P450 substrate. 2,3,5,6-Tetrafluorophenol was used in preparation of radioiodinated phenylalanine derivative which is useful in peptide synthesis, technetium-99m labeled antibodies.

Solubility
Partly miscible with water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.