Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5,6-Tetrafluorohydroquinone, 98%

$32.84 - $32.84

Chemical Identifiers

Chemical Name or Material 2,3,5,6-Tetrafluorohydroquinone
CAS 771-63-1
Molecular Formula C6H2F4O2
Formula Weight 182.08
MDL Number MFCD00002340
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1708803

  SDS
alfa aesar™
A1708803
1g Each for $32.84
Description & Specifications

Specifications

Formula Weight 182.08
Melting Point 168° to 170°C
Assay 98%
Chemical Name or Material 2,3,5,6-Tetrafluorohydroquinone
CAS 771-63-1
MDL Number MFCD00002340
Molecular Formula C6H2F4O2
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.