Please login to your online account to display your discounted pricing

2,3,5,6-Tetrafluorobenzoic acid, 98%, ACROS Organics™

$96.23 - $96.23

Chemical Identifiers

Chemical Name or Material 2,3,5,6-Tetrafluorobenzoic acid
Name Note 98%
CAS 652-18-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H2F4O2
Linear Formula HC6F4CO2H
Formula Weight 194.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC347370010

  SDS
thermo fisher scientific
347370010
Plastic bottle 1g Each for $96.23
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 194.08
Quantity 1g
Color White to almost white
TSCA TSCA
Melting Point 148° to 152°C
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye damage.
Assay Percent Range 98%
Linear Formula HC6F4CO2H
MDL Number MFCD00002408
Chemical Name or Material 2,3,5,6-Tetrafluorobenzoic acid
CAS 652-18-6
CAS Min % 97.5
Name Note 98%
Molecular Formula C7H2F4O2
Packaging Plastic bottle
Safety and Handling

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye damage.