missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,5,6-Tetrafluoro-1,4-benzenedimethanol, TCI America™

Manufacturer:  TCI America T273225G

 View more versions of this product

Catalog No. T273225G



Specifications

Specifications

2,3,5,6-Tetrafluoro-1,4-benzenedimethanol
129°C
92339-07-6
MFCD00229147
SDHKGYDQOGCLQM-UHFFFAOYSA-N
[2,3,5,6-tetrafluoro-4-(hydroxymethyl)phenyl]methanol
4098835
Crystal-Powder at 20°C
White-Yellow
25g
C8H6F4O2
2,3,5,6-Tetrafluoro-1,4-bis(hydroxymethyl)benzene, 2,3,5,6-Tetrafluoro-alpha,alpha′-dihydroxy-p-xylene, 2,3,5,6-Tetrafluoro-p-xylene-alpha,alpha′-diol
C(C1=C(C(=C(C(=C1F)F)CO)F)F)O
210.128
210.13
≥97.0% (GC)

Chemical Identifiers

92339-07-6
210.128
SDHKGYDQOGCLQM-UHFFFAOYSA-N
4098835
C(C1=C(C(=C(C(=C1F)F)CO)F)F)O
C8H6F4O2
MFCD00229147
2,3,5,6-Tetrafluoro-1,4-bis(hydroxymethyl)benzene, 2,3,5,6-Tetrafluoro-alpha,alpha′-dihydroxy-p-xylene, 2,3,5,6-Tetrafluoro-p-xylene-alpha,alpha′-diol
[2,3,5,6-tetrafluoro-4-(hydroxymethyl)phenyl]methanol
Safety and Handling

Safety and Handling

TSCA : No

Documents