Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,5,6-Tetrachloropyridine, 98%

Chemical Identifiers

Chemical Name or Material 2,3,5,6-Tetrachloropyridine
CAS 2402-79-1
Molecular Formula C5HCl4N
Formula Weight 216.88
MDL Number MFCD00092428
UN Number UN3077
Beilstein 129639
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1945803

  SDS
alfa aesar™
L1945803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1945806

  SDS
alfa aesar™
L1945806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1945814

  SDS
alfa aesar™
L1945814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 216.88
Chemical Name or Material 2,3,5,6-Tetrachloropyridine
MDL Number MFCD00092428
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA TSCA
Assay 98%
Molecular Formula C5HCl4N
CAS 2402-79-1
UN Number UN3077
Health Hazard 1 Danger
Flash Point 188°C (370°F)
Beilstein 129639
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 250° to 252°C
Melting Point 89° to 93°C
Quantity 1g
RTECS Number UT8225000
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.