Please login to your online account to display your discounted pricing

2,3,5,6-Tetrabromohydroquinone, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,3,5,6-Tetrabromohydroquinone
Name Note 98%
CAS 2641-89-6
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C6 H2 Br4 O2
Formula Weight 425.7
MDL Number MFCD00013970
Synonym 2,3,5,6-Tetrabromobenzene-1,4-diol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC335910250

  SDS
thermo fisher scientific
335910250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 425.7
Quantity 25g
TSCA TSCA
EINECS Number 220-142-9
CAS Max % 100.0
Synonym 2,3,5,6-Tetrabromobenzene-1,4-diol
CAS Min % 97.5
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
Name Note 98%
MDL Number MFCD00013970
CAS 2641-89-6
Chemical Name or Material 2,3,5,6-Tetrabromohydroquinone
Molecular Formula C6 H2 Br4 O2
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.